The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318865.1 | NP_001305794.1 | 5 - 6 | 675 | 101 |
NM_018960.5 | NP_061833.1 | 5 - 6 | 732 | 101 | |
NR_134899.1 | 4 - 5 | 589 | 101 | ||
GenBank mRNA | AF101477.1 | - | 5 - 6 | 728 | 101 |
BC032627.1 | - | 5 - 6 | 724 | 101 | |
FJ224320.1 | - | 5 - 6 | 728 | 101 | |
KU178603.1 | - | 5 - 6 | 718 | 101 | |
KU178604.1 | - | 5 - 6 | 661 | 101 | |
KU178605.1 | - | 4 - 5 | 575 | 101 | |
X62250.1 | - | 5 - 6 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318856.1 | NP_001305785.1 | 5 - 6 | 1104 | 101 |
NM_001318857.1 | NP_001305786.1 | 4 - 5 | 976 | 101 | |
NM_001318858.1 | NP_001305787.1 | 4 - 5 | 868 | 101 | |
NR_134890.1 | 4 - 5 | 930 | 101 | ||
NR_134891.1 | 3 - 4 | 833 | 101 | ||
NR_134892.1 | 4 - 5 | 961 | 101 | ||
GenBank mRNA | FY210916.1 | - | 4 - 5 | 509 | 101 |
FY213370.1 | - | 3 - 4 | 365 | 101 | |
FY213777.1 | - | 4 - 5 | 401 | 101 | |
FY213816.1 | - | 4 - 5 | 462 | 101 |