The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057161.3 | NP_476502.1 | 9 - 10 | 1305 | 87 |
NR_040101.1 | 8 - 9 | 1189 | 87 | ||
GenBank mRNA | AB055925.1 | - | 9 - 10 | 1169 | 87 |
AK097719.1 | - | 7 - 8 | 1031 | 87 | |
AK290384.1 | - | 9 - 10 | 1161 | 87 | |
AK302336.1 | - | 8 - 9 | 944 | 87 | |
AK308917.1 | - | 8 - 9 | 1311 | 87 | |
BC000295.2 | - | 9 - 10 | 1125 | 87 | |
BC001789.1 | - | 9 - 10 | 1140 | 87 | |
BC001793.2 | - | 9 - 10 | 1103 | 87 | |
BC007296.1 | - | 9 - 10 | 1155 | 87 | |
BC009460.1 | - | 9 - 10 | 1153 | 87 | |
BC021546.1 | - | 9 - 10 | 1142 | 87 | |
BC041793.1 | - | 8 - 9 | 1043 | 87 | |
BC045612.1 | - | 8 - 9 | 1043 | 87 |