Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEDD4
Entrez Gene ID
4734
Gene Name
neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase
Gene Aliases
NEDD4-1, RPF1
Chromosome Location
Chr.15: 55826917 - 55993746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284338.1 NP_001271267.1 12 - 13 3031 76
NM_001284339.1 NP_001271268.1 12 - 13 2983 76
NM_001284340.1 NP_001271269.1 11 - 12 2980 76
NM_006154.3 NP_006145.2 16 - 17 1665 76
NM_198400.3 NP_940682.2 9 - 10 2815 76
NR_104302.1 10 - 11 2918 76
XM_011521624.2 XP_011519926.1 15 - 16 1430 76
XM_011521625.2 XP_011519927.1 17 - 18 1603 76
XM_011521626.1 XP_011519928.1 15 - 16 1447 76
XM_011521627.1 XP_011519929.1 13 - 14 1270 76
GenBank mRNA AK304394.1 - 16 - 17 1531 76
AK307541.1 - 12 - 13 1282 76
AL832063.1 - 9 - 10 2816 76
AL832359.1 - 11 - 12 1503 76
AL833293.1 - 10 - 11 2256 76
AL833300.1 - 12 - 13 2013 76
AY550969.1 - 16 - 17 1544 76
BC035707.1 - 16 - 17 1509 76
BC136605.1 - 12 - 13 3008 76
BC144284.1 - 12 - 13 3008 76
BC144285.1 - 12 - 13 2960 76
BC152452.1 - 16 - 17 1556 76
BC152562.1 - 16 - 17 1556 76
D42055.1 - 16 - 17 1556 76

View Full Product Details