The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006154.3 | NP_006145.2 | 1 - 2 | 231 | 101 |
GenBank mRNA | AK304394.1 | - | 1 - 2 | 97 | 101 |
AY550969.1 | - | 1 - 2 | 110 | 101 | |
BC035707.1 | - | 1 - 2 | 75 | 101 | |
BC152452.1 | - | 1 - 2 | 122 | 101 | |
BC152562.1 | - | 1 - 2 | 122 | 101 | |
D42055.1 | - | 1 - 2 | 122 | 101 | |
DB451823.1 | - | 1 - 2 | 231 | 101 |