The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284338.1 | NP_001271267.1 | 16 - 17 | 3517 | 86 |
NM_001284339.1 | NP_001271268.1 | 16 - 17 | 3469 | 86 | |
NM_001284340.1 | NP_001271269.1 | 15 - 16 | 3466 | 86 | |
NM_006154.3 | NP_006145.2 | 20 - 21 | 2151 | 86 | |
NM_198400.3 | NP_940682.2 | 13 - 14 | 3301 | 86 | |
NR_104302.1 | 14 - 15 | 3404 | 86 | ||
XM_011521624.2 | XP_011519926.1 | 19 - 20 | 1916 | 86 | |
XM_011521625.2 | XP_011519927.1 | 21 - 22 | 2089 | 86 | |
XM_011521626.1 | XP_011519928.1 | 19 - 20 | 1933 | 86 | |
XM_011521627.1 | XP_011519929.1 | 17 - 18 | 1756 | 86 | |
GenBank mRNA | AK304394.1 | - | 20 - 21 | 2017 | 86 |
AK307541.1 | - | 16 - 17 | 1768 | 86 | |
AL832063.1 | - | 13 - 14 | 3302 | 86 | |
AL832359.1 | - | 15 - 16 | 1989 | 86 | |
AL833293.1 | - | 15 - 16 | 2742 | 86 | |
AL833300.1 | - | 16 - 17 | 2499 | 86 | |
AY550969.1 | - | 20 - 21 | 2030 | 86 | |
BC035707.1 | - | 20 - 21 | 1995 | 86 | |
BC136605.1 | - | 16 - 17 | 3494 | 86 | |
BC144284.1 | - | 16 - 17 | 3494 | 86 | |
BC144285.1 | - | 16 - 17 | 3446 | 86 | |
BC152452.1 | - | 20 - 21 | 2042 | 86 | |
BC152562.1 | - | 20 - 21 | 2042 | 86 | |
D42055.1 | - | 20 - 21 | 2042 | 86 |