Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF10
Entrez Gene ID
9639
Gene Name
Rho guanine nucleotide exchange factor 10
Gene Aliases
GEF10, SNCV
Chromosome Location
Chr.8: 1823976 - 1958641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308152.1 NP_001295081.1 17 - 18 2220 113
NM_001308153.1 NP_001295082.1 19 - 20 2423 113
NM_014629.3 NP_055444.2 18 - 19 2334 113
XM_005266041.3 XP_005266098.1 18 - 19 2276 113
XM_011534767.2 XP_011533069.1 16 - 17 2079 113
XM_011534768.1 XP_011533070.1 17 - 18 2199 113
XM_011534770.2 XP_011533072.1 17 - 18 2199 113
XM_017014003.1 XP_016869492.1 18 - 19 2239 113
GenBank mRNA AB002292.2 - 18 - 19 2333 113
AF009205.1 - 9 - 10 1168 113
BC036809.1 - 10 - 11 1884 113
BC112926.1 - 17 - 18 2208 113
CR749570.1 - 19 - 20 2384 113

View Full Product Details