The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171.5 | NP_001162.4 | 26 - 27 | 3772 | 71 |
XM_011522479.2 | XP_011520781.1 | 26 - 27 | 3883 | 71 | |
XM_011522480.1 | XP_011520782.1 | 26 - 27 | 3720 | 71 | |
XM_011522481.2 | XP_011520783.1 | 26 - 27 | 3919 | 71 | |
XM_017023212.1 | XP_016878701.1 | 25 - 26 | 3748 | 71 | |
XM_017023213.1 | XP_016878702.1 | 26 - 27 | 3806 | 71 | |
GenBank mRNA | AF076622.1 | - | 26 - 27 | 3758 | 71 |
AF168791.1 | - | 26 - 27 | 3735 | 71 | |
AM711638.1 | - | 24 - 25 | 3360 | 71 | |
AM774324.1 | - | 26 - 27 | 3735 | 71 | |
BC131732.1 | - | 26 - 27 | 3758 | 71 |