Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCN1
Entrez Gene ID
348980
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Gene Aliases
BCNG-1, BCNG1, EIEE24, HAC-2
Chromosome Location
Chr.5: 45254950 - 45696118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021072.3 NP_066550.2 5 - 6 1398 109
GenBank mRNA AF064876.1 - 5 - 6 1010 109

View Full Product Details