The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013386.4 | NP_037518.3 | 5 - 6 | 885 | 81 |
NM_213651.2 | NP_998816.1 | 5 - 6 | 796 | 81 | |
GenBank mRNA | AF123303.1 | - | 5 - 6 | 644 | 81 |
AJ619961.1 | - | 5 - 6 | 728 | 81 | |
AJ619987.1 | - | 5 - 6 | 674 | 81 | |
AK292567.1 | - | 5 - 6 | 796 | 81 | |
AK304165.1 | - | 5 - 6 | 879 | 81 | |
AK316304.1 | - | 5 - 6 | 839 | 81 | |
AK316506.1 | - | 5 - 6 | 707 | 81 | |
BC014519.1 | - | 5 - 6 | 741 | 81 | |
BC068561.1 | - | 5 - 6 | 774 | 81 |