The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002772.2 | NP_002763.2 | 13 - 14 | 1596 | 88 |
XM_011529654.1 | XP_011527956.1 | 15 - 16 | 1870 | 88 | |
XM_011529655.1 | XP_011527957.1 | 16 - 17 | 1788 | 88 | |
XM_011529656.2 | XP_011527958.1 | 16 - 17 | 1795 | 88 | |
XM_011529657.1 | XP_011527959.1 | 14 - 15 | 1825 | 88 | |
XM_011529658.1 | XP_011527960.1 | 13 - 14 | 1789 | 88 | |
XM_011529659.1 | XP_011527961.1 | 14 - 15 | 1760 | 88 | |
GenBank mRNA | BC111749.1 | - | 13 - 14 | 1585 | 88 |
U09860.1 | - | 13 - 14 | 1605 | 88 |