The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002772.2 | NP_002763.2 | 8 - 9 | 912 | 165 |
XM_011529654.1 | XP_011527956.1 | 10 - 11 | 1186 | 165 | |
XM_011529655.1 | XP_011527957.1 | 11 - 12 | 1104 | 165 | |
XM_011529656.2 | XP_011527958.1 | 11 - 12 | 1111 | 165 | |
XM_011529657.1 | XP_011527959.1 | 9 - 10 | 1141 | 165 | |
XM_011529658.1 | XP_011527960.1 | 8 - 9 | 1105 | 165 | |
XM_011529659.1 | XP_011527961.1 | 9 - 10 | 1076 | 165 | |
GenBank mRNA | BC111749.1 | - | 8 - 9 | 901 | 165 |
U09860.1 | - | 8 - 9 | 921 | 165 |