Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP23
Entrez Gene ID
84294
Gene Name
UTP23, small subunit processome component
Gene Aliases
C8orf53
Chromosome Location
Chr.8: 116766503 - 116774684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK055279.1 - 2 - 3 417 88

View Full Product Details