The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204144.2 | NP_001191073.1 | 1 - 2 | 270 | 85 |
NM_001321868.1 | NP_001308797.1 | 1 - 2 | 270 | 85 | |
NM_001321869.1 | NP_001308798.1 | 1 - 2 | 270 | 85 | |
NM_001321870.1 | NP_001308799.1 | 1 - 2 | 270 | 85 | |
NM_001321871.1 | NP_001308800.1 | 1 - 2 | 270 | 85 | |
NM_001321872.1 | NP_001308801.1 | 1 - 2 | 270 | 85 | |
NM_001321873.1 | NP_001308802.1 | 1 - 2 | 270 | 85 | |
XM_011515631.2 | XP_011513933.1 | 1 - 2 | 270 | 85 | |
GenBank mRNA | AK304298.1 | - | 1 - 2 | 263 | 85 |
DC333353.1 | - | 1 - 2 | 270 | 85 |