The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033604.1 | NP_001028776.1 | 1 - 2 | 508 | 89 |
NM_001033605.1 | NP_001028777.1 | 1 - 2 | 508 | 89 | |
NM_014451.3 | NP_055266.2 | 1 - 2 | 508 | 89 | |
NM_198428.2 | NP_940820.1 | 1 - 2 | 508 | 89 | |
XM_005249700.3 | XP_005249757.1 | 1 - 2 | 516 | 89 | |
XM_005249701.2 | XP_005249758.1 | 1 - 2 | 516 | 89 | |
XM_011515264.2 | XP_011513566.1 | 1 - 2 | 516 | 89 | |
XM_011515266.2 | XP_011513568.1 | 1 - 2 | 516 | 89 | |
XM_011515267.2 | XP_011513569.1 | 1 - 2 | 516 | 89 | |
XM_011515268.1 | XP_011513570.1 | 1 - 2 | 516 | 89 | |
XM_011515269.2 | XP_011513571.1 | 1 - 2 | 495 | 89 | |
XM_011515270.2 | XP_011513572.1 | 1 - 2 | 516 | 89 | |
XM_017011992.1 | XP_016867481.1 | 1 - 2 | 495 | 89 | |
XM_017011993.1 | XP_016867482.1 | 1 - 2 | 495 | 89 | |
XM_017011994.1 | XP_016867483.1 | 1 - 2 | 516 | 89 | |
GenBank mRNA | AK057660.1 | - | 1 - 2 | 486 | 89 |
AK314627.1 | - | 1 - 2 | 349 | 89 | |
BC103831.2 | - | 1 - 2 | 128 | 89 | |
BX647385.1 | - | 1 - 2 | 486 | 89 |