Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGA1
Entrez Gene ID
26088
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
Gene Aliases
-
Chromosome Location
Chr.22: 37607235 - 37633564 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001560.2 NP_001001560.1 13 - 14 1822 66
NM_001172687.1 NP_001166158.1 15 - 16 2071 66
NM_001172688.1 NP_001166159.1 16 - 17 2129 66
NM_013365.4 NP_037497.1 15 - 16 2083 66
XM_005261517.3 XP_005261574.1 16 - 17 2308 66
XM_005261518.1 XP_005261575.1 15 - 16 1768 66
XM_005261520.1 XP_005261577.1 15 - 16 1974 66
XM_005261522.1 XP_005261579.1 10 - 11 1325 66
XM_006724229.1 XP_006724292.1 16 - 17 2016 66
XM_011530122.1 XP_011528424.1 16 - 17 2241 66
XM_011530123.1 XP_011528425.1 15 - 16 1819 66
XM_011530124.1 XP_011528426.1 14 - 15 2047 66
XM_017028760.1 XP_016884249.1 16 - 17 2257 66
XM_017028761.1 XP_016884250.1 14 - 15 3424 66
XM_017028762.1 XP_016884251.1 14 - 15 1996 66
XM_017028763.1 XP_016884252.1 16 - 17 1943 66
XM_017028764.1 XP_016884253.1 13 - 14 1713 66
XM_017028765.1 XP_016884254.1 9 - 10 1258 66
GenBank mRNA AF190862.1 - 15 - 16 1711 66
AF218584.1 - 15 - 16 1700 66
AF233521.1 - 15 - 16 1718 66
AK075256.1 - 15 - 16 2061 66
AK092417.1 - 4 - 5 730 66
AK095427.1 - 8 - 9 973 66
AK122898.1 - 16 - 17 2129 66
AK225157.1 - 15 - 16 1705 66
AK290548.1 - 15 - 16 1722 66
AL110219.1 - 14 - 15 1607 66
BC000538.2 - 6 - 7 876 66
BC044629.1 - 13 - 14 1451 66
CR456493.1 - 15 - 16 1883 66

View Full Product Details