Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAG16
Entrez Gene ID
79582
Gene Name
sperm associated antigen 16
Gene Aliases
PF20, WDR29
Chromosome Location
Chr.2: 213284369 - 214410501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025436.2 NP_001020607.1 4 - 5 499 86
NM_024532.4 NP_078808.3 4 - 5 499 86
NR_047659.1 6 - 7 674 86
XM_006712746.2 XP_006712809.1 4 - 5 450 86
XM_011511814.1 XP_011510116.1 3 - 4 547 86
XM_011511815.1 XP_011510117.1 4 - 5 450 86
XM_011511816.2 XP_011510118.1 4 - 5 450 86
XM_011511817.1 XP_011510119.1 4 - 5 450 86
XM_011511818.1 XP_011510120.1 4 - 5 450 86
XM_011511819.2 XP_011510121.1 3 - 4 1198 86
XM_011511820.1 XP_011510122.1 4 - 5 450 86
XM_011511824.1 XP_011510126.1 4 - 5 450 86
XM_011511827.1 XP_011510129.1 4 - 5 450 86
XM_011511828.1 XP_011510130.1 4 - 5 450 86
XM_011511829.1 XP_011510131.1 4 - 5 450 86
XM_011511831.1 XP_011510133.1 4 - 5 450 86
XM_011511832.1 XP_011510134.1 4 - 5 450 86
XM_011511835.2 XP_011510137.1 3 - 4 542 86
XM_011511837.2 XP_011510139.1 4 - 5 450 86
XM_017004896.1 XP_016860385.1 5 - 6 579 86
XM_017004897.1 XP_016860386.1 2 - 3 3196 86
XM_017004899.1 XP_016860388.1 4 - 5 450 86
XM_017004900.1 XP_016860389.1 4 - 5 441 86
XM_017004901.1 XP_016860390.1 4 - 5 431 86
GenBank mRNA AF310672.1 - 4 - 5 436 86
AF426740.1 - 4 - 5 436 86
AF490390.1 - 4 - 5 429 86
AK026377.1 - 4 - 5 459 86
AK095036.1 - 3 - 4 547 86
AK310883.1 - 4 - 5 398 86
AL832962.1 - 4 - 5 388 86
AW207406.1 - 1 - 2 86
BC009614.1 - 4 - 5 460 86
BC067756.1 - 4 - 5 486 86
BC100282.1 - 4 - 5 447 86
CR457359.1 - 4 - 5 394 86

View Full Product Details