The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004599.3 | NP_004590.2 | 18 - 19 | 3398 | 155 |
NR_103834.1 | 21 - 22 | 3918 | 155 | ||
XM_006724310.2 | XP_006724373.1 | 18 - 19 | 3642 | 155 | |
XM_011530347.1 | XP_011528649.1 | 16 - 17 | 2852 | 155 | |
GenBank mRNA | AK299914.1 | - | 18 - 19 | 3227 | 155 |
AL834221.1 | - | 12 - 13 | 2177 | 155 | |
BC051385.1 | - | 21 - 22 | 3824 | 155 | |
BC051799.1 | - | 18 - 19 | 3364 | 155 | |
BC056158.1 | - | 18 - 19 | 3344 | 155 | |
CT841522.1 | - | 18 - 19 | 3288 | 155 | |
U02031.1 | - | 18 - 19 | 3323 | 155 |