The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004599.3 | NP_004590.2 | 1 - 2 | 281 | 136 |
NR_103834.1 | 1 - 2 | 281 | 136 | ||
XM_011530347.1 | XP_011528649.1 | 1 - 2 | 110 | 136 | |
XM_017028921.1 | XP_016884410.1 | 1 - 2 | 110 | 136 | |
XM_017028922.1 | XP_016884411.1 | 1 - 2 | 110 | 136 | |
GenBank mRNA | AK299914.1 | - | 1 - 2 | 110 | 136 |
BC051385.1 | - | 1 - 2 | 196 | 136 | |
BC051799.1 | - | 1 - 2 | 247 | 136 | |
BC056158.1 | - | 1 - 2 | 227 | 136 | |
CT841522.1 | - | 1 - 2 | 171 | 136 | |
U02031.1 | - | 1 - 2 | 206 | 136 |