Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IHH
Entrez Gene ID
3549
Gene Name
indian hedgehog
Gene Aliases
BDA1, HHG2
Chromosome Location
Chr.2: 219054420 - 219060516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002181.3 NP_002172.2 2 - 3 628 103
GenBank mRNA BC034757.1 - 2 - 3 298 103
BC136587.1 - 2 - 3 628 103
BC136588.1 - 2 - 3 628 103
L38517.1 - 1 - 2 283 103

View Full Product Details