Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D30
Entrez Gene ID
23329
Gene Name
TBC1 domain family member 30
Gene Aliases
-
Chromosome Location
Chr.12: 64759489 - 64881022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015279.1 NP_056094.1 11 - 12 1810 66
XM_017019085.1 XP_016874574.1 13 - 14 1691 66
XM_017019086.1 XP_016874575.1 13 - 14 1696 66
XM_017019087.1 XP_016874576.1 11 - 12 1562 66
GenBank mRNA AB023201.1 - 12 - 13 1405 66
AB449914.1 - 11 - 12 1502 66
AK055404.1 - 11 - 12 1752 66
AK299508.1 - 10 - 11 1589 66

View Full Product Details