Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENKUR
Entrez Gene ID
219670
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
C10orf63, CFAP106
Chromosome Location
Chr.10: 24981979 - 25062279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_072992.1 6 - 7 1153 67
GenBank mRNA BC026165.2 - 6 - 7 1155 67

View Full Product Details