Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFB4
Entrez Gene ID
149954
Gene Name
BPI fold containing family B member 4
Gene Aliases
C20orf186, LPLUNC4, RY2G5, dJ726C3.5
Chromosome Location
Chr.20: 33081512 - 33111751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182519.2 NP_872325.2 12 - 13 1655 115
GenBank mRNA AF549190.1 - 11 - 12 1523 115

View Full Product Details