Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNK7
Entrez Gene ID
10089
Gene Name
potassium two pore domain channel subfamily K member 7
Gene Aliases
K2p7.1, TWIK3
Chromosome Location
Chr.11: 65592855 - 65595996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033348.1 NP_203134.1 3 - 4 1016 109
GenBank mRNA AF110523.1 - 3 - 4 1016 109
BC103809.1 - 3 - 4 829 109

View Full Product Details