Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIB3
Entrez Gene ID
57761
Gene Name
tribbles pseudokinase 3
Gene Aliases
C20orf97, NIPK, SINK, SKIP3, TRB3
Chromosome Location
Chr.20: 380629 - 397559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021158.4 NP_066981.2 1 - 2 537 121
GenBank mRNA AJ697936.1 - 2 - 3 380 121
AK026945.1 - 2 - 3 290 121
AK222837.1 - 2 - 3 290 121
AY247738.1 - 1 - 2 536 121
BC019363.1 - 1 - 2 103 121
BC027484.1 - 1 - 2 290 121
BG828172.1 - 1 - 2 206 121

View Full Product Details