The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127615.1 | NP_001121087.1 | 12 - 13 | 1749 | 82 |
NM_001242863.1 | NP_001229792.1 | 12 - 13 | 2073 | 82 | |
NM_017993.3 | NP_060463.2 | 12 - 13 | 2099 | 82 | |
XM_005266439.3 | XP_005266496.1 | 12 - 13 | 2062 | 82 | |
XM_011535126.2 | XP_011533428.1 | 12 - 13 | 1799 | 82 | |
XM_011535127.2 | XP_011533429.1 | 13 - 14 | 2162 | 82 | |
XM_011535128.2 | XP_011533430.1 | 13 - 14 | 2112 | 82 | |
XM_011535132.2 | XP_011533434.1 | 11 - 12 | 1914 | 82 | |
XM_017020637.1 | XP_016876126.1 | 14 - 15 | 3381 | 82 | |
XM_017020638.1 | XP_016876127.1 | 17 - 18 | 2384 | 82 | |
XM_017020639.1 | XP_016876128.1 | 13 - 14 | 2110 | 82 | |
XM_017020640.1 | XP_016876129.1 | 11 - 12 | 1929 | 82 | |
GenBank mRNA | AK000956.1 | - | 11 - 12 | 1719 | 82 |
AY513282.1 | - | 9 - 10 | 1473 | 82 | |
BC024178.1 | - | 12 - 13 | 1759 | 82 | |
EF432052.1 | - | 9 - 10 | 1449 | 82 | |
KU178683.1 | - | 9 - 10 | 1449 | 82 | |
KU178684.1 | - | 8 - 9 | 1342 | 82 |