The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127615.1 | NP_001121087.1 | 7 - 8 | 889 | 96 |
NM_001242863.1 | NP_001229792.1 | 7 - 8 | 1213 | 96 | |
NM_017993.3 | NP_060463.2 | 7 - 8 | 1239 | 96 | |
XM_005266439.3 | XP_005266496.1 | 7 - 8 | 1202 | 96 | |
XM_011535126.2 | XP_011533428.1 | 7 - 8 | 939 | 96 | |
XM_011535127.2 | XP_011533429.1 | 8 - 9 | 1302 | 96 | |
XM_011535128.2 | XP_011533430.1 | 8 - 9 | 1252 | 96 | |
XM_011535132.2 | XP_011533434.1 | 6 - 7 | 1054 | 96 | |
XM_017020637.1 | XP_016876126.1 | 9 - 10 | 2521 | 96 | |
XM_017020638.1 | XP_016876127.1 | 12 - 13 | 1524 | 96 | |
XM_017020639.1 | XP_016876128.1 | 8 - 9 | 1250 | 96 | |
XM_017020640.1 | XP_016876129.1 | 6 - 7 | 1069 | 96 | |
XM_017020641.1 | XP_016876130.1 | 7 - 8 | 1202 | 96 | |
XM_017020642.1 | XP_016876131.1 | 7 - 8 | 1202 | 96 | |
GenBank mRNA | AK000956.1 | - | 6 - 7 | 859 | 96 |
AK299053.1 | - | 2 - 3 | 337 | 96 | |
AY513282.1 | - | 4 - 5 | 613 | 96 | |
BC024178.1 | - | 7 - 8 | 899 | 96 | |
EF432052.1 | - | 4 - 5 | 589 | 96 | |
KU178683.1 | - | 4 - 5 | 589 | 96 | |
KU178684.1 | - | 4 - 5 | 589 | 96 |