The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002213.4 | NP_002204.2 | 4 - 5 | 914 | 78 |
XM_005247436.2 | XP_005247493.1 | 4 - 5 | 1147 | 78 | |
XM_006713630.2 | XP_006713693.1 | 4 - 5 | 1147 | 78 | |
XM_017006351.1 | XP_016861840.1 | 4 - 5 | 685 | 78 | |
XM_017006352.1 | XP_016861841.1 | 4 - 5 | 624 | 78 | |
XM_017006353.1 | XP_016861842.1 | 4 - 5 | 656 | 78 | |
XM_017006354.1 | XP_016861843.1 | 4 - 5 | 1147 | 78 | |
XM_017006355.1 | XP_016861844.1 | 4 - 5 | 624 | 78 | |
GenBank mRNA | AK315429.1 | - | 4 - 5 | 650 | 78 |
BC006541.2 | - | 4 - 5 | 906 | 78 | |
J05633.1 | - | 4 - 5 | 655 | 78 | |
M35011.1 | - | 4 - 5 | 638 | 78 | |
X53002.1 | - | 4 - 5 | 945 | 78 |