The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175039.3 | NP_778204.1 | 2 - 3 | 287 | 60 |
GenBank mRNA | AB035172.1 | - | 1 - 2 | 32 | 60 |
BC036705.2 | - | 2 - 3 | 193 | 60 |