The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022900.4 | NP_075051.4 | 10 - 11 | 1580 | 85 |
XM_006716092.3 | XP_006716155.1 | 8 - 9 | 1075 | 85 | |
XM_011516495.2 | XP_011514797.1 | 10 - 11 | 1418 | 85 | |
XM_017012546.1 | XP_016868035.1 | 9 - 10 | 1203 | 85 | |
XM_017012547.1 | XP_016868036.1 | 10 - 11 | 1361 | 85 | |
GenBank mRNA | AB073397.1 | - | 2 - 3 | 353 | 85 |
AF355594.1 | - | 10 - 11 | 1373 | 85 | |
AF397424.2 | - | 10 - 11 | 1500 | 85 | |
AK025532.1 | - | 3 - 4 | 671 | 85 | |
AK123866.1 | - | 10 - 11 | 1418 | 85 | |
AK314329.1 | - | 10 - 11 | 1378 | 85 | |
BC063284.1 | - | 10 - 11 | 1565 | 85 |