The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022900.4 | NP_075051.4 | 12 - 13 | 1857 | 106 |
XM_006716092.3 | XP_006716155.1 | 10 - 11 | 1352 | 106 | |
XM_011516495.2 | XP_011514797.1 | 12 - 13 | 1695 | 106 | |
XM_017012546.1 | XP_016868035.1 | 11 - 12 | 1480 | 106 | |
XM_017012547.1 | XP_016868036.1 | 12 - 13 | 1638 | 106 | |
GenBank mRNA | AB073397.1 | - | 4 - 5 | 630 | 106 |
AF355594.1 | - | 12 - 13 | 1650 | 106 | |
AF397424.2 | - | 12 - 13 | 1777 | 106 | |
AK025532.1 | - | 5 - 6 | 107 | ||
AK123866.1 | - | 12 - 13 | 1695 | 106 | |
AK314329.1 | - | 12 - 13 | 1655 | 106 | |
BC063284.1 | - | 12 - 13 | 1842 | 106 |