The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022900.4 | NP_075051.4 | 14 - 15 | 2044 | 118 |
XM_006716092.3 | XP_006716155.1 | 12 - 13 | 1539 | 118 | |
XM_011516495.2 | XP_011514797.1 | 14 - 15 | 1882 | 118 | |
XM_017012546.1 | XP_016868035.1 | 13 - 14 | 1667 | 118 | |
XM_017012547.1 | XP_016868036.1 | 14 - 15 | 1825 | 118 | |
GenBank mRNA | AB073397.1 | - | 6 - 7 | 817 | 118 |
AF397424.2 | - | 14 - 15 | 1964 | 118 | |
AK024866.1 | - | 3 - 4 | 244 | 118 | |
AK025532.1 | - | 7 - 8 | 1136 | 118 | |
AK123866.1 | - | 14 - 15 | 1882 | 118 | |
AK314329.1 | - | 14 - 15 | 1842 | 118 | |
BC063284.1 | - | 14 - 15 | 2029 | 118 |