The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015130.2 | NP_055945.2 | 14 - 15 | 2711 | 129 |
GenBank mRNA | AB020689.2 | - | 14 - 15 | 2514 | 129 |
AB449892.1 | - | 14 - 15 | 2439 | 129 | |
AK127020.1 | - | 11 - 12 | 1999 | 129 | |
BC038804.1 | - | 5 - 6 | 825 | 129 | |
BC146758.1 | - | 14 - 15 | 2514 | 129 |