The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032382.4 | NP_115758.3 | 3 - 4 | 1482 | 75 |
GenBank mRNA | AK025968.1 | - | 1 - 2 | 604 | 75 |
AK056344.1 | - | 3 - 4 | 1470 | 75 | |
AK225391.1 | - | 1 - 2 | 604 | 75 | |
AK302605.1 | - | 3 - 4 | 1495 | 75 | |
BC017492.1 | - | 1 - 2 | 286 | 75 | |
BC121022.1 | - | 3 - 4 | 1442 | 75 | |
BC121023.1 | - | 3 - 4 | 1442 | 75 |