Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCL2
Entrez Gene ID
132954
Gene Name
phosducin like 2
Gene Aliases
GCPHLP
Chromosome Location
Chr.4: 55556523 - 55592275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152401.2 NP_689614.2 1 - 2 110 140
XM_005265728.3 XP_005265785.1 1 - 2 172 140
XM_017007745.1 XP_016863234.1 1 - 2 173 140
GenBank mRNA BC034431.1 - 1 - 2 110 140

View Full Product Details