The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258038.1 | NP_001244967.1 | 3 - 3 | 2228 | 103 |
NM_001258039.1 | NP_001244968.1 | 2 - 2 | 1972 | 103 | |
NM_005841.2 | NP_005832.1 | 2 - 2 | 2149 | 103 | |
NM_199327.2 | NP_955359.1 | 2 - 2 | 1982 | 103 | |
XM_005262686.1 | XP_005262743.1 | 3 - 3 | 2889 | 103 | |
XM_017007635.1 | XP_016863124.1 | 2 - 2 | 1962 | 103 | |
GenBank mRNA | AF041037.1 | - | 1 - 1 | 1214 | 103 |
AK026960.1 | - | 2 - 2 | 2126 | 103 | |
AY192146.1 | - | 2 - 2 | 1977 | 103 | |
AY590694.1 | - | 1 - 1 | 1780 | 103 | |
BC063856.1 | - | 2 - 2 | 2111 | 103 | |
CA447611.1 | - | 1 - 1 | 103 |