The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128429.2 | NP_001121901.1 | 20 - 21 | 2882 | 86 |
NM_001128430.1 | NP_001121902.1 | 20 - 21 | 2784 | 86 | |
NM_001254949.1 | NP_001241878.1 | 12 - 13 | 1698 | 86 | |
NM_020159.4 | NP_064544.2 | 20 - 21 | 2876 | 86 | |
NR_045644.1 | 21 - 22 | 2933 | 86 | ||
XM_017008463.1 | XP_016863952.1 | 20 - 21 | 2700 | 86 | |
XM_017008464.1 | XP_016863953.1 | 21 - 22 | 2883 | 86 | |
XM_017008465.1 | XP_016863954.1 | 21 - 22 | 2893 | 86 | |
GenBank mRNA | AB032948.2 | - | 19 - 20 | 2670 | 86 |
AK023990.1 | - | 5 - 6 | 665 | 86 | |
AK025302.1 | - | 15 - 16 | 1916 | 86 | |
AK301668.1 | - | 12 - 13 | 1698 | 86 | |
AL359929.1 | - | 11 - 12 | 1306 | 86 | |
AL512768.1 | - | 5 - 6 | 648 | 86 | |
AY008271.1 | - | 20 - 21 | 2679 | 86 | |
BC045534.1 | - | 20 - 21 | 2837 | 86 |