The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128429.2 | NP_001121901.1 | 21 - 22 | 3001 | 125 |
NM_001128430.1 | NP_001121902.1 | 21 - 22 | 2903 | 125 | |
NM_001254949.1 | NP_001241878.1 | 13 - 14 | 1817 | 125 | |
NM_020159.4 | NP_064544.2 | 21 - 22 | 2995 | 125 | |
NR_045644.1 | 22 - 23 | 3052 | 125 | ||
XM_017008463.1 | XP_016863952.1 | 21 - 22 | 2819 | 125 | |
XM_017008464.1 | XP_016863953.1 | 22 - 23 | 3002 | 125 | |
XM_017008465.1 | XP_016863954.1 | 22 - 23 | 3012 | 125 | |
GenBank mRNA | AB032948.2 | - | 20 - 21 | 2789 | 125 |
AK023990.1 | - | 6 - 7 | 784 | 125 | |
AK025302.1 | - | 16 - 17 | 2035 | 125 | |
AK301668.1 | - | 13 - 14 | 1817 | 125 | |
AL359929.1 | - | 12 - 13 | 1425 | 125 | |
AL512768.1 | - | 6 - 7 | 767 | 125 | |
AY008271.1 | - | 21 - 22 | 2798 | 125 | |
BC045534.1 | - | 21 - 22 | 2956 | 125 |