Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPIP5K2
Entrez Gene ID
23262
Gene Name
diphosphoinositol pentakisphosphate kinase 2
Gene Aliases
CFAP160, HISPPD1, IP7K2, VIP2
Chromosome Location
Chr.5: 103120254 - 103203523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276277.1 NP_001263206.1 22 - 23 3163 148
NM_001281471.1 NP_001268400.1 23 - 24 3208 148
NM_015216.3 NP_056031.2 21 - 22 2928 148
XM_005271935.1 XP_005271992.1 22 - 23 3162 148
XM_005271936.1 XP_005271993.1 22 - 23 3162 148
XM_005271938.1 XP_005271995.1 22 - 23 3162 148
XM_005277534.1 XP_005277591.1 23 - 24 3207 148
XM_005277535.1 XP_005277592.1 23 - 24 3204 148
XM_005277536.1 XP_005277593.1 23 - 24 3207 148
XM_005277538.1 XP_005277595.1 23 - 24 3207 148
XM_005277539.1 XP_005277596.1 23 - 24 3207 148
XM_005277541.1 XP_005277598.1 23 - 24 3207 148
XM_006714576.1 XP_006714639.1 23 - 24 3207 148
XM_006714577.1 XP_006714640.1 23 - 24 3207 148
XM_011543290.2 XP_011541592.1 23 - 24 3207 148
XM_017009269.1 XP_016864758.1 22 - 23 3159 148
XM_017009270.1 XP_016864759.1 22 - 23 3162 148
XM_017009271.1 XP_016864760.1 22 - 23 3159 148
XM_017009272.1 XP_016864761.1 22 - 23 3162 148
XM_017009273.1 XP_016864762.1 22 - 23 3159 148
XM_017009274.1 XP_016864763.1 22 - 23 3162 148
XM_017009275.1 XP_016864764.1 22 - 23 3159 148
GenBank mRNA AB007893.1 - 22 - 23 3153 148
AK025159.1 - 4 - 5 466 148
AK294787.1 - 23 - 24 3072 148
BC024591.1 - 21 - 22 2681 148

View Full Product Details