The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031714.3 | NP_001026884.3 | 1 - 2 | 131 | 57 |
NM_022489.3 | NP_071934.3 | 1 - 2 | 131 | 57 | |
NM_032714.2 | NP_116103.1 | 1 - 2 | 131 | 57 | |
GenBank mRNA | AK290083.1 | - | 1 - 2 | 116 | 57 |
BC006173.1 | - | 1 - 2 | 116 | 57 | |
BI489810.1 | - | 1 - 2 | 131 | 57 | |
BX248757.1 | - | 1 - 2 | 100 | 57 |