Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLG
Entrez Gene ID
5340
Gene Name
plasminogen
Gene Aliases
-
Chromosome Location
Chr.6: 160702193 - 160754054 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000301.3 NP_000292.1 6 - 7 776 89
GenBank mRNA AK312966.1 - 6 - 7 727 89
BC060513.1 - 6 - 7 702 89
M74220.1 - 6 - 7 713 89
X05199.1 - 6 - 7 718 89

View Full Product Details