Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STXBP5
Entrez Gene ID
134957
Gene Name
syntaxin binding protein 5
Gene Aliases
LGL3, LLGL3, Nbla04300
Chromosome Location
Chr.6: 147204184 - 147390476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127715.2 NP_001121187.1 27 - 28 3584 93
NM_139244.4 NP_640337.3 25 - 26 3476 93
XM_006715339.2 XP_006715402.1 26 - 27 3710 93
XM_011535453.1 XP_011533755.1 26 - 27 3308 93
XM_017010267.1 XP_016865756.1 22 - 23 3655 93
GenBank mRNA AB073398.1 - 16 - 17 2315 93
AK055484.1 - 8 - 9 1294 93
AK090549.1 - 17 - 18 2294 93
AK128121.1 - 26 - 27 3303 93
AL834152.1 - 28 - 29 3633 93
BC113382.1 - 25 - 26 3349 93
BC113408.1 - 25 - 26 3349 93

View Full Product Details