Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANGEL1
Entrez Gene ID
23357
Gene Name
angel homolog 1
Gene Aliases
Ccr4e, KIAA0759
Chromosome Location
Chr.14: 76786473 - 76812947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015305.3 NP_056120.2 3 - 4 988 65
XM_017021115.1 XP_016876604.1 3 - 4 995 65
GenBank mRNA AB018302.1 - 3 - 4 885 65
AL137268.1 - 3 - 4 886 65
BC028714.2 - 3 - 4 935 65

View Full Product Details