The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286408.1 | NP_001273337.1 | 6 - 7 | 1152 | 154 |
NM_001286409.1 | NP_001273338.1 | 3 - 4 | 857 | 154 | |
NM_198081.4 | NP_932347.2 | 7 - 8 | 1367 | 154 | |
XM_011535703.2 | XP_011534005.1 | 7 - 8 | 1265 | 154 | |
XM_011535704.2 | XP_011534006.1 | 7 - 8 | 1143 | 154 | |
XM_011535706.1 | XP_011534008.1 | 6 - 7 | 1109 | 154 | |
XM_011535709.2 | XP_011534011.1 | 3 - 4 | 1299 | 154 | |
XM_011535710.2 | XP_011534012.1 | 5 - 6 | 976 | 154 | |
XM_017010677.1 | XP_016866166.1 | 7 - 8 | 1434 | 154 | |
XM_017010678.1 | XP_016866167.1 | 7 - 8 | 1347 | 154 | |
XM_017010679.1 | XP_016866168.1 | 7 - 8 | 1245 | 154 | |
XM_017010680.1 | XP_016866169.1 | 7 - 8 | 1313 | 154 | |
XM_017010681.1 | XP_016866170.1 | 7 - 8 | 1223 | 154 | |
XM_017010683.1 | XP_016866172.1 | 6 - 7 | 1106 | 154 | |
XM_017010685.1 | XP_016866174.1 | 6 - 7 | 1233 | 154 | |
XM_017010686.1 | XP_016866175.1 | 6 - 7 | 1218 | 154 | |
XM_017010687.1 | XP_016866176.1 | 6 - 7 | 1144 | 154 | |
GenBank mRNA | AK093571.1 | - | 6 - 7 | 1152 | 154 |