The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308237.1 | NP_001295166.1 | 3 - 4 | 361 | 86 |
NM_015534.5 | NP_056349.1 | 3 - 4 | 269 | 86 | |
XM_005270725.3 | XP_005270782.1 | 3 - 4 | 344 | 86 | |
XM_005270727.3 | XP_005270784.1 | 3 - 4 | 344 | 86 | |
XM_017000971.1 | XP_016856460.1 | 3 - 4 | 344 | 86 | |
XM_017000972.1 | XP_016856461.1 | 3 - 4 | 332 | 86 | |
GenBank mRNA | AK310514.1 | - | 3 - 4 | 266 | 86 |
BC035079.2 | - | 3 - 4 | 353 | 86 | |
BX641001.1 | - | 3 - 4 | 270 | 86 | |
BX641096.1 | - | 3 - 4 | 255 | 86 | |
DA722345.1 | - | 3 - 4 | 361 | 86 |