The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172818.1 | NP_001166289.1 | 1 - 2 | 548 | 151 |
GenBank mRNA | AK300100.1 | - | 1 - 2 | 534 | 151 |