The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024646.2 | NP_078922.1 | 10 - 11 | 1890 | 132 |
XM_006710898.3 | XP_006710961.1 | 10 - 11 | 1777 | 132 | |
GenBank mRNA | AB051517.1 | - | 10 - 11 | 1840 | 132 |
AK056645.1 | - | 8 - 9 | 834 | 132 | |
AK095643.1 | - | 9 - 10 | 1530 | 132 | |
BC029832.1 | - | 8 - 9 | 904 | 132 | |
BX647643.1 | - | 8 - 9 | 1063 | 132 |