The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001856.3 | NP_001847.3 | 14 - 15 | 1485 | 92 |
XM_005270481.1 | XP_005270538.1 | 14 - 15 | 1480 | 92 | |
XM_011540722.1 | XP_011539024.1 | 14 - 15 | 1480 | 92 | |
XM_011540723.1 | XP_011539025.1 | 14 - 15 | 1480 | 92 | |
XM_011540724.1 | XP_011539026.1 | 14 - 15 | 1480 | 92 | |
XM_011540726.2 | XP_011539028.1 | 11 - 12 | 996 | 92 | |
XM_011540727.2 | XP_011539029.1 | 4 - 5 | 241 | 92 | |
XM_011540729.1 | XP_011539031.1 | 14 - 15 | 1480 | 92 | |
XM_011540730.1 | XP_011539032.1 | 14 - 15 | 1480 | 92 | |
XM_017000338.1 | XP_016855827.1 | 14 - 15 | 1480 | 92 | |
XM_017000339.1 | XP_016855828.1 | 14 - 15 | 1480 | 92 | |
GenBank mRNA | AB209571.1 | - | 10 - 11 | 792 | 92 |
M92642.1 | - | 15 - 16 | 1273 | 92 |