Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
Chromosome Location
Chr.1: 31652247 - 31704167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001856.3 NP_001847.3 24 - 25 2027 56
XM_005270481.1 XP_005270538.1 24 - 25 2022 56
XM_011540722.1 XP_011539024.1 24 - 25 2022 56
XM_011540723.1 XP_011539025.1 24 - 25 2022 56
XM_011540724.1 XP_011539026.1 24 - 25 2022 56
XM_011540726.2 XP_011539028.1 21 - 22 1538 56
XM_011540727.2 XP_011539029.1 14 - 15 783 56
XM_011540728.2 XP_011539030.1 2 - 3 265 56
XM_011540729.1 XP_011539031.1 24 - 25 2022 56
XM_011540730.1 XP_011539032.1 24 - 25 2022 56
XM_017000338.1 XP_016855827.1 24 - 25 2022 56
GenBank mRNA AB209571.1 - 20 - 21 1334 56
M92642.1 - 25 - 26 1812 56
S57132.1 - 7 - 8 408 56
X14963.1 - 8 - 9 456 56
X15038.1 - 8 - 9 456 56

View Full Product Details