Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
Chromosome Location
Chr.1: 31652247 - 31704167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001856.3 NP_001847.3 27 - 28 2208 100
XM_005270481.1 XP_005270538.1 27 - 28 2203 100
XM_011540722.1 XP_011539024.1 27 - 28 2203 100
XM_011540723.1 XP_011539025.1 27 - 28 2203 100
XM_011540724.1 XP_011539026.1 27 - 28 2203 100
XM_011540726.2 XP_011539028.1 24 - 25 1719 100
XM_011540727.2 XP_011539029.1 17 - 18 964 100
XM_011540728.2 XP_011539030.1 5 - 6 446 100
XM_011540729.1 XP_011539031.1 27 - 28 2203 100
XM_011540730.1 XP_011539032.1 27 - 28 2203 100
XM_017000338.1 XP_016855827.1 27 - 28 2203 100
XM_017000339.1 XP_016855828.1 26 - 27 2167 100
XM_017000340.1 XP_016855829.1 4 - 5 409 100
GenBank mRNA AB209571.1 - 23 - 24 1515 100
M92642.1 - 28 - 29 1993 100
S57132.1 - 10 - 11 589 100
X14963.1 - 11 - 12 637 100
X15038.1 - 11 - 12 637 100

View Full Product Details