Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
Chromosome Location
Chr.1: 31652247 - 31704167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001856.3 NP_001847.3 34 - 35 2749 136
XM_005270481.1 XP_005270538.1 34 - 35 2744 136
XM_011540722.1 XP_011539024.1 34 - 35 2744 136
XM_011540723.1 XP_011539025.1 34 - 35 2744 136
XM_011540724.1 XP_011539026.1 33 - 34 2621 136
XM_011540726.2 XP_011539028.1 31 - 32 2260 136
XM_011540727.2 XP_011539029.1 24 - 25 1505 136
XM_011540728.2 XP_011539030.1 12 - 13 987 136
XM_011540729.1 XP_011539031.1 34 - 35 2744 136
XM_011540730.1 XP_011539032.1 34 - 35 2744 136
XM_017000338.1 XP_016855827.1 34 - 35 2744 136
XM_017000339.1 XP_016855828.1 33 - 34 2708 136
XM_017000340.1 XP_016855829.1 11 - 12 950 136
XM_017000341.1 XP_016855830.1 11 - 12 740 136
GenBank mRNA AB209571.1 - 30 - 31 2056 136
M92642.1 - 35 - 36 2534 136
S57132.1 - 17 - 18 1130 136
X14963.1 - 18 - 19 1178 136
X15038.1 - 18 - 19 1178 136

View Full Product Details